Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300301

Sigma-Aldrich

Anti-SULT1B1 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-1B, Anti-Cytosolic, Anti-Member 1, Anti-Sulfotransferase 1B2, Anti-Sulfotransferase family

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULT1B1(27284)

General description

Cytosolic sulfotransferase (ST) enzymes catalyze the sulfate conjugation of many drugs, xenobiotic compounds, hormones, and neurotransmitters. Sulfotransferases such as SULT1B1 catalyze the biotransformation of a large number of endogenous compounds such as neurotransmitters, steroids, bile acids, and thyroid hormones, as well as drugs and xenobiotics. SULT1B1 contains a binding site for the sulfate donor, 3-prime-phosphoadenosine 5-prime-phosphosulfate, and a cysteine residue conserved in the ST1 gene family of sulfotransferases. SULT1B1 shares between 51.5% and 56.3% amino acid sequence identity with members of the phenol sulfotransferase gene family. Northern blot analysis detects 4 SULT1B1 transcripts between 79 kb and 13 kb expressed at highest levels in liver, peripheral blood leukocytes, colon, small intestine, and spleen. Lower expression is detected in lung, placenta, and thymus, and no expression is detected in other tissues. Recombinant rat and human SULT1B1, expressed in E. coli, catalyze sulfation of p-nitrophenol, 3,3-prime,5-triiodothyronine (T3), and dopamine, but not of beta-estradiol and dehydroepiandrosterone. SULT1B1 shows higher affinities for formation of T3 sulfate than do the phenol sulfotransferases ST1A3 (SULT1A1) or ST1A5. Bacterially-expressed SULT1B1 sulfates small phenols such as 1-naphthol and p-nitrophenol and thyroid hormones, including 3,3-prime-diiodothyronine, triiodothyronine, reverse triiodothyronine, and thyroxine. No activity is detected against several sterols or dopamine.

Immunogen

SULT1B1 (NP_055280, 1-30)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human SULT1B1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service