Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300244

Sigma-Aldrich

Anti-rde-1 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Hypothetical protein rde-1, Anti-RNA interference promoting factor RDE- 1, Anti-Rde-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Caenorhabditis elegans

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

General description

Double-stranded (ds) RNA is a potent sequence-specific inhibitor of gene function. RNA interference (RNAi) is a cellular defense mechanism that uses double-stranded RNA (dsRNA) as a sequence-specific trigger to guide the degradation of homologous single-stranded RNAs. RNAi is a multistep process involving several proteins and at least one type of RNA intermediate, a population of small 21–25 nt RNAs (called siRNAs) that are initially derived from cleavage of the dsRNA trigger. Genetic studies have implicated several RNA interference-deficient (rde) family members in germline maintenance and development, and several simple loss of function mutants have been identified. Family members rde-1 and rde-4 are required for RNAi but are not essential for organismal viability. While rde-1 and rde-4 are distinct from other RNAi-deficient family members both both for their inability to mobilize transposons and lack of chromosome loss, each appears to have a distinct role in the interference mechanism. Evidence indicates that rde-4 is involved before or during production of siRNAs, whereas rde-1 acts after the siRNAs have been formed.

Immunogen

RDE (NP_741611, )
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of C. elegans Rde-1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service