Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300243

Sigma-Aldrich

Anti-drh-1 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Dicer related helicase protein 1, Anti-Dicer-related helicase, Anti-Drh-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Caenorhabditis elegans

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Caenorhabditis elegans ... Drh-1(177425)

General description

Human Drh-1 protein possesses an RNA helicase motif containing a DEXH box in its amino terminus and an RNA motif in the carboxy terminus. DICER, also known as helicase-MOI, is required by the RNA interference and small temporal RNA (stRNA) pathways to produce the active small RNA component that represses gene expression. The 21-nucleotide small temporal RNA (stRNA) let7 regulates developmental timing in C. elegans and probably in other bilateral animals. In Drosophila, a developmentally regulated precursor RNA is cleaved by an RNA interference-like mechanism to produce mature let7 stRNA. Targeted disruption in cultured human cells of the mRNA encoding the enzyme Drh-1, which acts in the RNA interference pathway, leads to accumulation of the LET7 precursor. Thus, the RNA interference and stRNA pathways intersect. Both pathways require the RNA processing enzyme Drh-1 to produce the active small RNA component that represses gene expression.

Immunogen

DRH (NP_501018, 12-46)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of C. elegans Drh-1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia D Consalvo et al.
eLife, 13 (2024-05-15)
Invertebrates use the endoribonuclease Dicer to cleave viral dsRNA during antiviral defense, while vertebrates use RIG-I-like Receptors (RLRs), which bind viral dsRNA to trigger an interferon response. While some invertebrate Dicers act alone during antiviral defense, Caenorhabditis elegans Dicer acts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service