Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300242

Sigma-Aldrich

Anti-dcr-1 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Dcr-1, Anti-Endoribonuclease dcr-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Caenorhabditis elegans

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Caenorhabditis elegans ... Dcr-1(176138)

General description

The Dcr-1 gene encodes a bidentate ribonuclease that is homologous to E. coli RNAse III.
DCR-1 (dicer 1, ribonuclease III) is an RNase ΙΙΙ endoribonuclease. It is located on human chromosome 14q32.

Immunogen

DCR (NP_498761, 1008-1042)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the central region of C. elegans Dcr-1.

Biochem/physiol Actions

Dcr-1 is required both for RNA interference and for the synthesis of small developmental RNAs. Dcr-1 interacts in vivo with RDE-4, a double-stranded RNA (dsRNA) binding protein required for RNAi that interacts with trigger dsRNAs and may function to deliver dsRNAs to Dcr-1 for endonucleolytic processing.
DCR-1 (dicer 1, ribonuclease III) has the ability to cleave dsRNA into smaller fragments that mediate RNAi. It plays an important role in miRNA processing and participates in innate immunity. DCR-1 is essential for the production of endo-siRNAs implicated in gene silencing and epigenetic modulation.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new role for DCR-1/DICER in C. elegans innate immunity against the highly virulent bacterium Bacillus thuringiensis DB27
Iatsenko, et al.
Infection and Immunity, IAI-00700 (2013)
FOXE1 polymorphisms are associated with familial and sporadic nonmedullary thyroid cancer susceptibility
Tomaz, et al.
Clinical Endocrinology, 77(6), 926-933 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service