Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300226

Sigma-Aldrich

Anti-IL29 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Cytokine ZCYTO21, Anti-IFN-λ-1, Anti-IL-29, Anti-Interferon λ-1, Anti-Interleukin-29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL29(282618)

General description

IFN-λ-1,-2, and -3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family of interferons. All three IFN-λs use a distinct receptor system composed of an IFN-λR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defense similar but distinct than the type I Interferons.

Immunogen

IL29 (NP_742152, 58-95)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human L29.

Biochem/physiol Actions

Interleukin-29 (IL-29) enhances the production of interleukins from monocytes. It has been observed that it stimulates antiviral responses to herpes simplex virus 1 in vitro. Serums from patients with rheumatoid arthritis have high levels of IL29.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Wu et al.
Autoimmunity, 46(1), 40-43 (2012-10-09)
Interferon (IFN)-λ1 is a newly described cytokine that is known for its proinflammatory activity in viral infection and in cancer. Because recent studies indicated that IFN-λ can influence significantly the innate and adaptive immune response, we studied IFN-λ in a
Fang Wang et al.
Arthritis research & therapy, 14(5), R228-R228 (2012-10-20)
The immunoregulatory function of interleukin (IL)-29 has recently been recognized. However, little is known about the involvement of IL-29 in the pathogenesis of rheumatoid arthritis (RA). This study aimed to examine the expression profiles of IL-29 in blood, synovial fluid
Sergei V Kotenko et al.
Nature immunology, 4(1), 69-77 (2002-12-17)
We report here the identification of a ligand-receptor system that, upon engagement, leads to the establishment of an antiviral state. Three closely positioned genes on human chromosome 19 encode distinct but paralogous proteins, which we designate interferon-lambda1 (IFN-lambda1), IFN-lambda2 and
K Lopušná et al.
Acta virologica, 58(4), 325-332 (2014-12-19)
Lambda interferons (IFN-λ) are known to induce potent antiviral response in a wide variety of target cells. They activate the same intracellular signalling pathways and have similar biological activities as IFN-α/β, including antiviral activity, but signal via distinct receptor complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service