Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300157

Sigma-Aldrich

Anti-HRX (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HRX(4297)

General description

The gene variously symbolized ALL1, HRX, or MLL located on 11q23 has been demonstrated to be fused with a number of translocation partners in cases of leukemia. Tse et al. (1995) characterized 2 t(1;11)(q21;q23) translocations that fused the MLL gene to a gene on chromosomal band 1q21, AF1Q, in 2 infants with acute myelomonocytic leukemia. In one of these patients, the derivative chromosome 11 represented an in-frame fusion of the N-terminal portion of the MLL gene to the complete AF1Q open reading frame, whereas the derivative chromosome 1 did not give rise to an open reading frame. This observation suggested that the N-terminal portion of the MLL gene is critical for leukemogenesis in translocations involving band 11q23.

Immunogen

HRX (HRX_HUMAN, 2591-2625)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human HRX.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service