Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300152

Sigma-Aldrich

Anti-MOZ/MYST3 (N-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-Histone acetyltransferase MYST3, Anti-MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3, Anti-MYST protein 3, Monocytic leukemia zinc finger protein, Runt-related transcription factor-binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYST3(7994)

General description

MOZ (monocytic leukemia zinc finger protein) is also known as lysine acetyltransferase 6 (KAT6A) and belongs to the MYST (MOZ, YBF2/SAS3, SAS2 and TIP60) family. The KAT6 gene is mapped to human chromosomes 8p11.21. The structure of KAT6 is evolutionarily conserved in organisms ranging from yeast to mammals. The encoded protein contains a nuclear localization domain (including H15), a histone-acetyltransferase domain (HAT), an acidic glutamate/aspartate-rich region, a double-plant homeodomain finger that interacts with acetylated histone H3 tails (PHD 1 and 2) and a serine and methionine-rich region containing a transactivation domain. In humans, the gene is broadly expressed in different tissues, strongly in the brain and low in the heart.
MOZ/MYST3 is a histone acetyltransferase which may act as a transcriptional coactivator for RUNX1 and RUNX2.

Immunogen

MOZ/MYST3 (NP_001092882, 72-108)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human MYST3.

Biochem/physiol Actions

KAT6A (lysine acetyltransferase 6A) is a histone acetyltransferase. It is a transcriptional co-activator. It acetylates histone H3 at K14 and K9 and H4 at K5, K8, K12 and K16 in vitro. KAT6A is associated with the regulation of cell cycle and stem cell homeostasis. In acute myeloid leukemia the involvement of KAT6A along with CREB binding protein has been observed. KAT6A acts as an oncogene. Mutations and abnormal regulation in KAT6 gene is associated with solid tumors and developmental disorders in human. It serves as a key epigenetic regulator of hematopoiesis. Mutations in the gene lead to abnormal acetylation of K9 and K18 of histone3 as well as changes in the P53 signaling. Therefore, affecting multiple cellular processes and controlling disease conditions.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fu Huang et al.
Molecular and cellular biology, 36(14), 1900-1907 (2016-05-18)
The lysine acetyltransferase 6 (KAT6) histone acetyltransferase (HAT) complexes are highly conserved from yeast to higher organisms. They acetylate histone H3 and other nonhistone substrates and are involved in cell cycle regulation and stem cell maintenance. In addition, the human
Valerie A Arboleda et al.
American journal of human genetics, 96(3), 498-506 (2015-03-03)
Chromatin remodeling through histone acetyltransferase (HAT) and histone deactylase (HDAC) enzymes affects fundamental cellular processes including the cell-cycle, cell differentiation, metabolism, and apoptosis. Nonsense mutations in genes that are involved in histone acetylation and deacetylation result in multiple congenital anomalies
Emma Tham et al.
American journal of human genetics, 96(3), 507-513 (2015-03-03)
Through a multi-center collaboration study, we here report six individuals from five unrelated families, with mutations in KAT6A/MOZ detected by whole-exome sequencing. All five different de novo heterozygous truncating mutations were located in the C-terminal transactivation domain of KAT6A: NM_001099412.1:
Xiaozhe Xiong et al.
Nature chemical biology, 12(12), 1111-1118 (2016-10-25)
Recognition of histone covalent modifications by 'reader' modules constitutes a major mechanism for epigenetic regulation. A recent upsurge of newly discovered histone lysine acylations, such as crotonylation (Kcr), butyrylation (Kbu), and propionylation (Kpr), greatly expands the coding potential of histone
Brittany Turner-Ivey et al.
Neoplasia (New York, N.Y.), 16(8), 644-655 (2014-09-16)
The chromosome 8p11-p12 amplicon is present in 12% to 15% of breast cancers, resulting in an increase in copy number and expression of several chromatin modifiers in these tumors, including KAT6A. Previous analyses in SUM-52 breast cancer cells showed amplification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service