Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300134

Sigma-Aldrich

Monoclonal Anti-SET07 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-H4-K20-HMTase, Anti-Histone H4-K20 methyltransferase, Anti-PR/SET07, PR/SET domain-containing protein 07, SET domain-containing protein 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

43AT890, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SET07(387893)

Related Categories

General description

SET07 is a histone methyltransferase that methylates Lys-20 of histone H4. H4 Lys-20 methylation represents a specific tag for epigenetic transcriptional repression. The nuclear SET07 protein, which associates with silent chromatin on euchromatic arms but shows no association with constitutive heterochromatin, prefers nucleosomes as substrate compared to free histones. It appears that SET07 may play a role in maintaining silent chromatin by preventing neighboring acetylation of H4 tail. Although the SET domain contains the active site of enzymatic activity, both sequences upstream and downstream of the SET domain are required for methyltransferase activity.

Immunogen

SET07 (AAF97812, N Terminal)
This antibody was raised using purified recombinant GST fusion protein encoding N-terminal of human SET07.

Physical form

Purified monoclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service