Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300126

Sigma-Aldrich

Anti-NEK2 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HsPK 21, Anti-HsPK21, Anti-NEK 2, Anti-NEK 2A, Anti-Serine threonine protein kinase Nek2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEK2(4751)

General description

NEK2 is closely related in its catalytic domain to the serine/threonine protein kinase NIMA of Aspergillus nidulans which is required for entry into mitosis and may function in parallel to the universal mitotic inducer p34cdc2. Like NIMA, the Nek2 protein is almost undetectable during G1 but accumulates progressively throughout S, reaching maximal levels in late G2. These observations demonstrate that NEK2 resembles Aspergillus NIMA, both in its catalytic domain, and cell cycle-dependent expression. Recombinant NEK2 is active as a serine/threonine-specific protein kinase and may undergo autophosphorylation. Both human NEK2 and fungal NIMA phosphorylate a similar, but not identical, set of proteins and synthetic peptides. NEK2 is shown to be expressed most abundantly in adult testis and there is increasing evidence that NEK2 is abnormally expressed in a wide variety of human cancers.

Immunogen

NEK2 (Q96QN9, 355-392)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human NEK2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service