Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300119

Sigma-Aldrich

Anti-PI4K II (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Phosphatidylinositol 4-kinase type II, Anti-Type II α phosphatidylinositol 4-kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PI4K II(55361)

General description

Phosphatidylinositolpolyphosphates (PtdInsPs) are centrally involved in many biologic processes, ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4KII phosphorylates PtdIns at the D-4 position, an essential step in the biosynthesis of PtdInsPs. PI4K II is activated by detergent and inhibited by adenosine. Overexpression of PI4KII in COS-7 cells increases synthesis of PtdIns4P. Some cells overexpressing PI4KII have scattered or no perinuclear Golgi. Knockdown of PI4KII by RNA interference (RNAi) does not disrupt the Golgi, and some cells show expanded Golgi. RNAi reduces the Golgi level of PtdIns4P and blocks the association between AP1 and the trans-Golgi network. PI4KII RNAi had little effect on intra-Golgi trafficking, but it inhibited export to plasma membrane export by 35%. It has been proposed that PI4KII generates PtdIns4P-rich domains within the Golgi that specify docking of the AP1 coat machinery.

Immunogen

PI4K II (NP_060895, 1-30)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human PI4K II.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service