Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1300104

Sigma-Aldrich

Anti-CAMK2δ-like (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CAMK2G, Anti-CaM-kinase II γ chain, Anti-Calcium/calmodulin-dependent protein kinase type II γ chain, CaM kinase II γ subunit, CaMK-II γ subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK2(818)

General description

Ca2+/calmodulin-dependent kinase II γ (CaMKIIγ) gene is mapped to human chromosome 10q22.2. The encoded protein is a multifunctional serine/threonine kinase and is specific to various substrates. CaMKII contains four isoforms: CaMKIIα/β/γ/δ. CaMKII is ubiquitously expressed in humans. The protein consists of an N-terminal catalytic domain, a regulatory domain, and a C-terminal association domain.
CaM-kinase II (CAMK2) is a prominent Ser/Thr protein kinase in the central nervous system that may function in long-term potentiation and neurotransmitter release. Likely autophosphorylation of Thr-286 allows the kinase to switch from a calmodulin-dependent to a calmodulin-independent state. CAMK2 is composed of four different chains: alpha, beta, gamma, and delta. The different isoforms assemble into homo- or heteromultimeric holoenzymes composed of 8 to 12 subunits.

The previously assigned protein identifier O15378 has been merged into Q13555. Full details can be found on the UniProt database.

Immunogen

CAMK2 (NP_751911, 314-348)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-term region of human CAMK2 delta-like.

Biochem/physiol Actions

CaMKIIγ (Ca2+/calmodulin-dependent kinase II γ) is mostly expressed in tumor-initiating cells. CaMKIIγ regulates the progression of cancer cells in leukemia, lung cancer, and liver cancer. CaMKIIγ participates in the β-catenin, Stat3 (signal transducer and activator of transcription 3), and NF-κB (nuclear factor kappa-light-chain-enhancer of activated B cells) signal pathways..

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas J Hund et al.
Trends in cardiovascular medicine, 25(5), 392-397 (2015-01-13)
Protein phosphorylation is a central mechanism in vertebrates for the regulation of signaling. With regard to the cardiovascular system, phosphorylation of myocyte targets is critical for the regulation of excitation contraction coupling, metabolism, intracellular calcium regulation, mitochondrial activity, transcriptional regulation
Lam C Tsoi et al.
Nature communications, 6, 7001-7001 (2015-05-06)
Psoriasis is a chronic autoimmune disease with complex genetic architecture. Previous genome-wide association studies (GWAS) and a recent meta-analysis using Immunochip data have uncovered 36 susceptibility loci. Here, we extend our previous meta-analysis of European ancestry by refined genotype calling
Shoujie Chai et al.
Oncotarget, 6(18), 16069-16083 (2015-05-13)
Highly tumorigenic stem-like cells, considered tumor-initiating cells (TICs), are the main cause of lung cancer initiation, relapse, and drug resistance. In this study, we identified that Ca2+/calmodulin-dependent protein kinase IIγ (CaMKIIγ) was aberrantly expressed in highly tumorigenic stem-like lung cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service