Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300031

Sigma-Aldrich

Anti-Mouse Dnmt3l antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DNA (cytosine-5)-methyltransferase 3-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Dnmt3L(54427)

General description

mDnmt3L (DNA methyltransferase 3L) does not appear to be catalytically active due to loss of the critical active site residues. It may function not directly as a DNA methyltransferase but rather as a regulator of methylation at imprinted loci. It is required specifically for the establishment of genomic imprints but is dispensable for their propagation. It is essential for the de novo methylation of single-copy DNA sequences. The protein is well-express in testis, thymus, ovary, and heart. mDnmt3L, which belongs to the 5-cytosine methyltransferase family, contains 1 ADD-type zinc finger.

Immunogen

DNMT3L (Q9CWR8, 393-431)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the mouse Dnmt3L sequence within the last 100 aa. The antibody may recognize the human homolog (44 kDa).

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service