Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S9450

Sigma-Aldrich

Anti-Sam68 (N-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Gap-associated tyrosine phosphoprotein, 62-KD, Anti-KH domain-containing, RNA-binding, signal transduction-associated protein 1, Anti-KHDR1, Anti-KHDRBS1, Anti-Src-associated in mitosis 68 kDa protein, Anti-p68

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

form

buffered aqueous solution

mol wt

antigen ~68 kDa

technique(s)

western blot: 1:500-1:1000 using using K562 lys

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KHDRBS1(10657)

General description

Src-associated in mitosis 68 kDa protein (Sam68/KHDR1) or K homology (KH) domain-containing, RNA binding, signal transduction associated 1 (KHDRBS1) is a member of the signal transducer and activator of RNA (STAR) family of RNA-binding proteins. This protein contains a KH (hnRNP K homology) RNA-binding domain located within a larger domain of 200 amino acids with RNA binding activity named GSG (GRP33/Sam68/GLD-1) domain. The Sam68 gene is mapped to human chromosome 1p35.

Specificity

Anti-Sam68 (N-terminal) recognizes human Sam68.

Application

Anti-Sam68 (N-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Src-associated in mitosis 68 kDa protein (Sam68/KHDR1/KHDRBS1) participates in signal transduction, RNA metabolism, apoptosis, transcription, cell cycle, and regulation. Besides binding to RNA and its modulator proteins, Sam68 contains proline-rich motifs and multiple potential tyrosine phosphorylation sites that are involved in the binding to proteins with Src homology 3 (SH3) and SH2 domains. It controls the activation of nuclear transcription factor kappa B (NF-κB).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing is not recommended. Storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuyu Chen et al.
Molecular medicine reports, 20(4), 3728-3734 (2019-09-06)
Hyperglycemia promotes podocyte apoptosis and contributes to the pathogenesis of diabetic nephropathy (DN). However, the mechanisms of hyperglycemia‑induced podocyte apoptosis remain unknown. Recent studies have implicated Src‑associated substrate during mitosis of 68 kDa (Sam68) in various cellular processes including RNA metabolism
Nilva K Cervigne et al.
Human molecular genetics, 23(10), 2618-2628 (2014-01-10)
A significant proportion (up to 62%) of oral squamous cell carcinomas (OSCCs) may arise from oral potential malignant lesions (OPMLs), such as leukoplakia. Patient outcomes may thus be improved through detection of lesions at a risk for malignant transformation, by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service