Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S9321

Sigma-Aldrich

Saikosaponin D from Bupleurum falcatnum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H68O13
Molecular Weight:
780.98
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥95% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC1(C)CC[C@]23[C@H](O)C[C@@]4(C)[C@]5(C)CC[C@@]6([H])[C@@](CO)(C)[C@@H](O[C@]7([H])O[C@H](C)[C@H](O)[C@H](O[C@]8([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)[C@H]7O)CC[C@]6(C)[C@@]5([H])C=C[C@]4(OC3)[C@]2([H])C1

InChI

1S/C42H68O13/c1-21-28(46)33(55-34-31(49)30(48)29(47)22(18-43)53-34)32(50)35(52-21)54-27-10-11-37(4)23(38(27,5)19-44)8-12-39(6)24(37)9-13-42-25-16-36(2,3)14-15-41(25,20-51-42)26(45)17-40(39,42)7/h9,13,21-35,43-50H,8,10-12,14-20H2,1-7H3/t21-,22-,23-,24-,25-,26-,27+,28+,29-,30+,31-,32-,33+,34+,35+,37+,38+,39-,40+,41-,42+/m1/s1

InChI key

KYWSCMDFVARMPN-LCSVLAELSA-N

Application

Saikosaponins such as Saikosaponin D are used to help analyse and resolve the activities of herbal medicines such as Sho-saiko-to.

Biochem/physiol Actions

Saikosaponin D inhibits cell growth of human cancer cells by inducing apoptosis and blocking cell cycle progression in the G1 phase. Saikosaponin D was found to stimulate corticotropin-releasing factor (CRF) gene expression and CRF release.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Sakaida et al.
Journal of hepatology, 28(2), 298-306 (1998-03-26)
A herbal medicine, Sho-saiko-to (TJ-9), has recently been orally administered to patients with chronic liver disease in Japan and has been reported to inhibit the development of hepatocellular carcinoma. The aim of this study was to investigate whether TJ-9 has
K Kayano et al.
Journal of hepatology, 29(4), 642-649 (1998-11-21)
It is of extreme importance to prevent liver fibrosis and subsequent progression to liver cirrhosis. The aim of our study was to elucidate in vitro whether Sho-saiko-to exerted inhibitory effects on hepatic stellate cells. Hepatic stellate cells were isolated from
Sho-saiko-to: Japanese herbal medicine for protection against hepatic fibrosis and carcinoma.
Shimizu I.
Journal of Gastroenterology and Hepatology, 15, Suppl D84-Suppl D90 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service