Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S9314

Sigma-Aldrich

Sesamin

≥95%, crystalline

Synonym(s):

Fagarol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O6
CAS Number:
Molecular Weight:
354.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Sesamum indicum (sesame)

assay

≥95%

form

crystalline

shipped in

wet ice

storage temp.

−20°C

SMILES string

C1Oc2ccc(cc2O1)[C@H]3OC[C@H]4[C@@H]3CO[C@@H]4c5ccc6OCOc6c5

InChI

1S/C20H18O6/c1-3-15-17(25-9-23-15)5-11(1)19-13-7-22-20(14(13)8-21-19)12-2-4-16-18(6-12)26-10-24-16/h1-6,13-14,19-20H,7-10H2/t13-,14-,19+,20+/m0/s1

InChI key

PEYUIKBAABKQKQ-AFHBHXEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sesamin is a fat-soluble furfuran lignan present in Sesamum indicum L seeds.[1]

Application

Sesamin has been used:
  • as a lignan to study its effects on the osteoblastic differentiation of rat bone marrow stromal cells (BMSCs) [2]
  • as a standard for the isolation of sesamin from sesame oil [3]
  • as an enterolignan to study its effects on the proliferation of estrogen-receptor (ER)-positive human breast cancer MCF-7 cells [4]

Biochem/physiol Actions

Non-competitive Δ5-desaturase inhibitor.
Sesamin can lower cholesterol effects in humans. It is involved in preventing high blood pressure and increases vitamin E levels.[5] Sesamin exhibits anti-cancer, anti-proliferative, anti-metastatic and anti-inflammatory properties. It also exhibits pro-apoptotic and pro-autophagocytic properties. Sesamin has its effects on lipid and carbohydrate metabolism. It also has effects on hypertension and free radical scavenging.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong-Ping Ma et al.
Medical science monitor : international medical journal of experimental and clinical research, 25, 5312-5320 (2019-07-18)
BACKGROUND Osteoporosis is a common osteopathy, resulting in fractures, especially in elder people. Sesamin has many pharmacological effects, including supplying calcium. However, how sesamin might prevent osteoporosis is still under study. MATERIAL AND METHODS Bone marrow stromal cells (BMSCs) extracted
Amin F Majdalawieh et al.
European journal of pharmacology, 815, 512-521 (2017-10-17)
Sesamin is the major active ingredient is Sesamum indicum seeds. Several studies revealed that sesamin possesses potent anti-cancer properties. The anti-cancer effects of sesamin have been mainly attributed to its anti-proliferative, pro-apoptotic, anti-inflammatory, anti-metastatic, anti- and pro-angiogenic, and pro-autophagocytic activities.
N Hirose et al.
Journal of lipid research, 32(4), 629-638 (1991-04-01)
The effects of sesamin, a lignan from sesame oil, on various aspects of cholesterol metabolism were examined in rats maintained on various dietary regimens. When given at a dietary level of 0.5% for 4 weeks, sesamin reduced the concentration of
Value addition in sesame: A perspective on bioactive components for enhancing utility and profitability
Pathak N, et al.
Pharmacognosy reviews, 8(16), 147-147 (2014)
Jean Fotie et al.
Journal of natural products, 70(10), 1650-1653 (2007-09-21)
Three benzo[ c]phenanthridine alkaloids have been isolated from the stem bark of Garcinia lucida: dihydrochelerythrine ( 1), 6-acetonyldihydrochelerythrine ( 2), and its new derivative, ( S)1''-(9,10-dihydro-2',3'-dihydroxy-7,8-dimethoxy-10-methyl-1,2-benzophenanthridin-9-yl)propan-2''-one (lucidamine A) ( 3). The new diisoprenylated derivative of lucidamine B ( 4) was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service