S9188
STAT1-α/β (699-709)
>98% (HPLC), solid
Synonym(s):
Signal Transducer and Activator of Transcription 1-α/β Fragment 699-709 human, mouse
About This Item
assay
>98% (HPLC)
form
solid
UniProt accession no.
storage temp.
−20°C
InChI
1S/C56H94N12O18/c1-11-29(7)44(67-50(79)38(24-33-16-18-34(72)19-17-33)60-40(73)25-59-52(81)42(58)31(9)70)53(82)61-35(15-13-14-22-57)48(77)68-46(32(10)71)55(84)62-36(20-21-41(74)75)47(76)63-37(23-27(3)4)49(78)66-45(30(8)12-2)54(83)64-39(26-69)51(80)65-43(28(5)6)56(85)86/h16-19,27-32,35-39,42-46,69-72H,11-15,20-26,57-58H2,1-10H3,(H,59,81)(H,60,73)(H,61,82)(H,62,84)(H,63,76)(H,64,83)(H,65,80)(H,66,78)(H,67,79)(H,68,77)(H,74,75)(H,85,86)/t29-,30-,31+,32+,35-,36-,37-,38-,39-,42-,43-,44-,45-,46-/m0/s1
InChI key
KTUYWMXWXJIBIH-KCGLDMGFSA-N
Gene Information
human ... STAT1(6772)
Amino Acid Sequence
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service