Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S8945

Sigma-Aldrich

Anti-Syntaxin 8 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27 kDa

species reactivity

rat

technique(s)

immunoprecipitation (IP): 10-20 μg using NRK cell lysate
microarray: suitable
western blot: 2-4 μg/mL using whole cell extract of the rat kidney NRK cell line and the rat pheochromocytoma PC12 cell line

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STX8(9482)
mouse ... Stx8(55943)
rat ... Stx8(59074)

General description

Syntaxin 8 (Stx8) is a member of the soluble NSF (N-ethyl-maleimide-sensitive protein) attachment proteins receptor (t-SNARE) family. Syntaxin 8 shares 23% identity with syntaxin 6 and is ubiquitously expressed in multiple tissues, with highest levels in the heart. This protein is anchored to membranes by a C-terminal hydrophobic domain and appears to contain a helical domain. Stx8 is localized to membrane compartments of the early endosome (EE). This protein has two unique di-leucine-based motifs. Stx8 gene is mapped to human chromosome 17p12.

Specificity

Anti-Syntaxin 8 specifically recognizes rat syntaxin 8 (27 kDa).

Immunogen

synthetic peptide corresponding to amino acids 117-134 located at the mid-region of rat syntaxin 8, conjugated to KLH. This sequence is identical in mouse syntaxin 8 and highly conserved (two amino acid substitutions) in human syntaxin 8.

Application

Anti-Syntaxin 8 antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Syntaxin 8 (Stx8) protein has been suggested to function in clathrin-independent vesicular transport and membrane fusion events necessary for protein transport from the early endosome (EE) to the late endosome (LE). This protein may also participate in vesicular trafficking and docking.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntaxin 8 has two functionally distinct di-leucine-based motifs
Kasai K, et al.
Cellular and Molecular Biology Letters, 13(1), 144-154 (2008)
V Thoreau et al.
Biochemical and biophysical research communications, 257(2), 577-583 (1999-04-13)
We report the cloning of a cDNA encoding human syntaxin 8 (STX8), using the regulator (R) domain of the cystic fibrosis transmembrane conductance regulator (CFTR) as a bait to screen a human fetal lung cDNA library by the yeast two-hybrid
Andoni Lancha et al.
Archives of medical research, 46(1), 47-53 (2014-12-20)
Obesity is associated with increased adipose tissue inflammation as well as with the development of type 2 diabetes (T2D). Syntaxin 8 (STX8) is a protein required for the transport of endosomes. In this study we analyzed the relationship of STX8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service