Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7296

Sigma-Aldrich

Derivatized Silica Gel Octyl (C8) Zorbax® LP 100/40 C8

40 μm, average diameter 100 Å

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41115700

packaging

pkg of 10 g

surface area

380 m2/g

matrix

fully porous particle

particle size

40 μm

pore size

100 Å average diameter

operating pH

2-8

application(s)

food and beverages

separation technique

reversed phase

InChI

1S/C10H23ClSi.O2Si/c1-4-5-6-7-8-9-10-12(2,3)11;1-3-2/h4-10H2,1-3H3;

InChI key

MBMCIQRWRYDPRE-UHFFFAOYSA-N

Application

Recommended for separation of hydrophobic peptides and proteins, including membrane proteins.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Zorbax is a registered trademark of Agilent Technologies, Inc.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service