Key Documents
S6894
Sigma-Aldrich
Sigma lab markers
Fine tip, red
About This Item
UNSPSC Code:
41103032
NACRES:
NB.35
color
red
General description
Writes on cold, wet surfaces. Quick-drying. Alcohol based permanent marker removed by alcohol.
Choose from one of the most recent versions:
Certificates of Analysis (COA)