Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6884

Sigma-Aldrich

Sera from baboon

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

biological source

baboon

form

lyophilized powder

storage temp.

2-8°C

General description

An esterase called pregnancy-associated esterase was found in baboon sera that preferentially hydrolyzes α-naphthyl butyrate [1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Cheng et al.
The Journal of experimental zoology, 241(1), 113-121 (1987-01-01)
Baboon serum samples were resolved by starch gel electrophoresis and polyacrylamide gradient gel electrophoresis and stained with naphthol substrates for esterase activity. An esterase that hydrolyzed alpha-naphthyl butyrate in preference to alpha-naphthyl acetate was found in very high activities in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service