Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6823

Sigma-Aldrich

SGK3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CISK, SGKL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

66-90 nmol/min·mg

mol wt

~82 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SGK3(23678)

Biochem/physiol Actions

SGK3 is a member of the SGK family and encodes a phosphoprotein with a PX (phox homology) domain. PKD1 can phosphorylate and activate SGK3 in vitro. A 10-fold increase in PKD1 increases the phosphorylation status of SGK3. When expressed in human embryonic kidney cells, IGF1 and peroxide significantly activate SGK3, and the activation could be reduced by preincubation with inhibitors of PI3 kinase. A yeast 2-hybrid screen found direct interaction between human SGK3 and GSK3b. SGK3 phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Gamper et al.
Pflugers Archiv : European journal of physiology, 445(1), 60-66 (2002-10-25)
The serum- and glucocorticoid-dependent kinase SGK1 was originally identified as a glucocorticoid-sensitive gene. Subsequently, the two homologous kinases SGK2 and SGK3 have been cloned, being products of distinct genes, which are differentially expressed and share 80% identity in amino acid
T Kobayashi et al.
The Biochemical journal, 344 Pt 1, 189-197 (1999-11-05)
The catalytic domain of serum- and glucocorticoid-induced protein kinase (SGK) is 54% identical with protein kinase B (PKB) and, like PKB, is activated in vitro by 3-phosphoinositide-dependent protein kinase-1 (PDK1) and in vivo in response to signals that activate phosphatidylinositol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service