Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6695

Sigma-Aldrich

Anti-Sin3A, C-Terminal antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 160 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μg using 293-T cell lysates
microarray: suitable
western blot: 2.5-5 μg/mL using HeLa nuclear extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIN3A(25942)
mouse ... Sin3a(20466)
rat ... Sin3a(363067)

General description

Human Sin3A is a 1273 amino acids protein containing paired amphipathic helix (PAH) domains, important for protein-protein interactions.
SIN3A (switch-insensitive 3 family member A) is a transcriptional repressor and MeCP2 (methyl-CpG binding protein 2) interactor. It is located on human chromosome 15q24. The product encoded by SIN3A gene is a constituent of the histone deacetylase (HDAC) complex.

Immunogen

synthetic peptide corresponding to amino acids 1267-1282 of human Sin3A, conjugated to KLH via an N-terminal added cysteine residue. The immunizing peptide is conserved in human, mouse, and rat and is not present in Sin3B.

Application

Anti-Sin3A, C-Terminal antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

SIN3A (switch-insensitive 3 family member A) along Sin3B is essential for transcription and growth suppressor functions of the mitotic spindle assembly checkpoint protein (Mad1) and interferon-induced GTP-binding protein (Mx1). It also controls cellular functions like differentiation, oncogenesis, apoptosis and cell fate determination. The Sin3A/HDAC corepressor comprises Sin3A along with histone deacetylases (HDAC1 and 2), pRB-associated proteins (RbAp46, RbAp48) and Sin3A Associated Protein 30 h(SAP30). Repression of transcription by the Sin3A/HDAC complex interaction with O-GlcNAc transferase (OGT) leads to the inactivation of transcription factors and RNA Polymerase II.
SIN3A (switch-insensitive 3 family member A) plays an important role in the growth of early embryos. It helps in the multiplication and survival of various cells by suppressing diverse signaling pathways. It acts as a transcriptional regulator for the formation of cortical brain. Absence of Sin3A can suppress the development of breast cancer cell by elevating apoptosis.

Physical form

Solution in 0.01 M phosphate buffered saline containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haploinsufficiency of MeCP2-interacting transcriptional co-repressor SIN3A causes mild intellectual disability by affecting the development of cortical integrity
Witteveen JS, et al.
Nature Genetics, 48(8), 877-887 (2016)
Maximum growth and survival of estrogen receptor-alpha positive breast cancer cells requires the Sin3A transcriptional repressor
Ellison-Zelski SJ and Alarid ET
Molecular Cancer, 9 (2010)
Identification and characterization of three new components of the mSin3A corepressor complex
Fleischer TC, et al.
Molecular and Cellular Biology, 23(10), 3456-3467 (2003)
Decreased expression of the SIN3A gene, a candidate tumor suppressor located at the prevalent allelic loss region 15q23 in non-small cell lung cancer
Suzuki H, et al.
Lung Cancer, 59(1), 24-31 (2008)
Recruitment of O-GlcNAc transferase to promoters by corepressor mSin3A: coupling protein O-GlcNAcylation to transcriptional repression
Yang X, et al.
Cell, 110(1), 69-80 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service