Key Documents
S6440
Nunc® Lab-Tek® II - CC2 Chamber Slide system
1 well
Synonym(s):
chamber slides, microscope slides, microscopy slides
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
colorless
polystyrene chamber
sterility
sterile
feature
CE marked
manufacturer/tradename
Nunc 154739
packaging
pkg of carton of 16
W × L
25 mm × 75 mm
well area
8.6 cm2
wells
1
suitability
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
General description
Nunc® Lab-Tek® II - CC2 Chamber Slide System
  • Chemically coated growth surface on glass slide that mimics polylysine
  • Provides binding sites optimal for fastidious cells (e.g. neurons)
  • Growth surface remains stable without refrigeration
  • Light blue frosted writing area
  • Slide seperator included in each pack
  • CE marked
Legal Information
CC2 is a trademark of Thermo Fisher Scientific or its subsidiaries
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
comparable product
Choose from one of the most recent versions:
Certificates of Analysis (COA)