Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

S6071

Sigma-Aldrich

Anti-Synphilin-1 (C-terminal) antibody produced in rabbit

enhanced validation

~2 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SNCAIP, Anti-Synuclein-α-interacting protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 130 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~2 mg/mL

technique(s)

western blot: 0.25-0.5 μg/mL using HEK-293 cells expressing human synphilin-1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SNCAIP(9627)
mouse ... Sncaip(67847)
rat ... Sncaip(307309)

General description

Synphilin-1 (synuclein a-interacting protein 1, SNCAIP), is a cytoplasmic protein with six ankyrin-like repeats, coiled-coil domain, and an ATP/GTP binding domain.[1] It is highly expressed in brain, heart, placenta[2] and presynaptic nerve terminals.[3] The synphilin gene produces at least nine transcript variants encoding seven distinct proteins.

Immunogen

synthetic peptide corresponding to amino acids 911-919 of human synphilin-1, conjugated to KLH. This sequence is identical in several species including mouse, rat, and dog synphilin-1.

Application

Anti-Synphilin-1 (C-terminal) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Cells with this mutation are more prone to staurosporine-induced cell death highlighting its cytoprotective functionality.[3] Synphilin-1 associates with and is ubiquitinated by several proteins, including α -synuclein, parkin, dorfin and siah E3 ubiquitin protein ligase 1 (SIAH1),[4] and is a major component of Lewy bodies in Parkinson′s disease (PD).[3] In synaptic vesicles it is modulated by α-synuclein.[3] The central domain of synphilin-1 has been suggested to be required for the formation of aggregates and cytotoxicity.[5] Synphilin-1 association with α -synuclein promotes the formation of cytosolic inclusions.[6] In human postmortem brain tissue, synphilin-1, like a-synuclein is present in neurophil.[7] Mutation of synphilin-1 in PD patients at amino acid R621C, is implicated in the sequestration, ubiquitination and proteasomal inhibition leading to increased accumulation of toxic intermediates.[3]

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service