S6024
Spacers for vertical standard gel, 16.5 cm wide
size 1.5 mm × 28 cm
All Photos(1)
About This Item
UNSPSC Code:
12352200
size
1.5 mm × 28 cm
Packaging
Set includes two spacers. Rounded corner mates with gasket and bottom corners of rectangular glass plate. Gasket design eliminates the need for a bottom spacer.
Choose from one of the most recent versions:
Certificates of Analysis (COA)