Key Documents
S5769
Sigma-Aldrich
Sigma lab markers
Extra-Fine tip, black
About This Item
UNSPSC Code:
41181600
color
black
General description
Writes on cold, wet surfaces. Quick-drying. Alcohol based permanent marker removed by alcohol.
Choose from one of the most recent versions:
Certificates of Analysis (COA)