Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5683

Sigma-Aldrich

Anti-STAT1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 91 kDa

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): 4 μg using STAT1 from 0.5–1 mg of human Raji and Jurkat, and mouse spleen cell lysates
western blot (chemiluminescent): 1 μg/mL using human Raji, Jurkat, and Hut, or mouse CTLL and spleen cell lysates

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT1(6772)
mouse ... Stat1(20846)

Specificity

The antibody is specific for STAT1.

Immunogen

GST fusion protein containing the C-terminal of human STAT1, amino acids 714-750, expressed in Escherichia coli.

Physical form

Solution in 0.07 M Tris-glycine, pH 7.4, containing 0.105 M NaCl, 0.035% sodium azide, and 30% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuijie Liu et al.
Oncotarget, 8(17), 29101-29115 (2017-04-19)
Among the diverse co-regulatory relationships between transcription factors (TFs) and microRNAs (miRNAs), feedback loops have received the most extensive research attention. The co-regulation of TFs and miRNAs plays an important role in colorectal cancer (CRC) growth. Here, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service