S5547
Anti-Syntaxin 3 antibody produced in rabbit
affinity isolated antibody, lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
rat
technique(s)
western blot (chemiluminescent): 1:200-1:300
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... STX3(6809)
mouse ... Stx3(20908)
rat ... Stx3(81802)
Immunogen
synthetic peptide corresponding to amino acids 2-16 of mouse, rat, or human syntaxin 3 (with additional C-terminal cysteine). The epitope is specific for alternative splicing forms of syntaxin 3 and is identical in mouse, rat, and human.
Physical form
Lyophilized at 0.3 mg/mL from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 0.025% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hugo Lachuer et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(8), e2207425120-e2207425120 (2023-02-18)
Lysosomal exocytosis is involved in many key cellular processes but its spatiotemporal regulation is poorly known. Using total internal reflection fluorescence microscopy (TIRFM) and spatial statistics, we observed that lysosomal exocytosis is not random at the adhesive part of the
Vasundhara Kandachar et al.
Journal of cell science, 131(24) (2018-11-09)
The Arf4-rhodopsin complex (mediated by the VxPx motif in rhodopsin) initiates expansion of vertebrate rod photoreceptor cilia-derived light-sensing organelles through stepwise assembly of a conserved trafficking network. Here, we examine its role in the sorting of VAMP7 (also known as
Taishin Takuma et al.
FEBS letters, 587(6), 583-589 (2013-02-06)
The exocytosis of salivary proteins is mainly regulated by cAMP, although soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNAREs), which mediate cAMP-dependent exocytic membrane fusion, have remained unidentified. Here we examined the effect of isoproterenol (ISO) and cytochalasin D (CyD) on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service