Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S4946

Sigma-Aldrich

Anti-Superoxide Dismutase (EC-SOD) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Extracellular SOD, Anti-SOD3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 31:35 kDa (doublet)

species reactivity

mouse

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extract of mouse lung and a chemiluminescent detection reagent

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Sod3(20657)

General description

Superoxide Dismutase (SOD) or CuZn-SOD (SOD1), a cytoplasmic and mitochondrial intermembrane space protein, is located on human chromosome 21q22. It belongs to superoxide dismutase multigene family.

Immunogen

synthetic peptide corresponding to amino acid residues 1-19 of mouse EC-SOD with a C-terminal added cysteine, conjugated to KLH.

Application

Anti-Superoxide Dismutase (EC-SOD) antibody has been used in immunofluorescent staining, western blot analysis{58,59} and immunohistochemical studies.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Superoxide Dismutase (SOD) or CuZn-SOD (SOD1) mutations results in amyotrophic lateral sclerosis. It acts as a mediator of the HMF (hypomagnetic field) effect.

Physical form

Solution in 0.01 M phosphate buffered saline containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiac myocyte-specific transgenic ecSOD targets mitochondria to protect against Ca2+ induced permeability transition
Luo J, et al.
Frontiers in Physiology, 4 (2013)
Low intensity radiofrequency radiation:a new oxidant for living cells
Yakymenko I, et al.
Scientific periodical, 3(1), 1-3 (2014)
Debaprasad Koner et al.
Fish physiology and biochemistry, 47(6), 1909-1932 (2021-10-06)
The deduced amino acid sequences from the complete cDNA coding sequences of three antioxidant enzyme genes (sod1, sod2, and cat) demonstrated that phylogenetically the magur catfish (Clarias magur) is very much close to other bony fishes with complete conservation of
Jianzhu Luo et al.
Frontiers in physiology, 4, 295-295 (2013-11-07)
ecSOD function has prototypically been associated with the extracellular space due to its secretion and localization to the extracellular matrix. A myocyte-specific ecSOD transgenic mouse has shown that it can also be localized to the myocyte intracellular compartment and is
A review of asthma genetics: gene expression studies and recent candidates
Malerba G1 and Pignatti PF
Journal of applied genetics, 46(1), 93-104 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service