S4697
Monoclonal Anti-Seladin-1 antibody produced in mouse
~2 mg/mL, clone SL-14, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-24-dehydro-cholesterol reductase (DCHR24), Anti-Diminuto/Dwarf-1 homolog, Anti-Selective Alzheimer’s disease indicator-1
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
SL-14, monoclonal
form
buffered aqueous solution
mol wt
antigen ~60 kDa
species reactivity
human
packaging
antibody small pack of 25 μL
concentration
~2 mg/mL
technique(s)
immunocytochemistry: suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using extracts of HEK-293T cells expressing human seladin-1
isotype
IgG2b
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... DHCR24(1718)
General description
Immunogen
Application
- enzyme linked immunosorbent assay (ELISA),
- immunoblotting
- immunocytochemistry
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service