S4637
Sera from rhesus monkey
lyophilized powder (from clotted whole blood)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
biological source
rhesus monkey
form
lyophilized powder (from clotted whole blood)
storage temp.
2-8°C
Application
Monkey serum has been used to study ageing properties associated with the inhibition of fibroblast migration. Not tested for use in cell culture.
Physical form
0.2m filtered prior to lyophilization from the indicated amount
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H Kondo et al.
Mechanisms of ageing and development, 79(2-3), 141-150 (1995-04-14)
In order to determine whether serum modified cellular aging in vivo, we previously studied the effects of serum from various mammals of different ages on cell functions such as proliferation and migration, and reported that cell migration was more greatly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service