Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S4191

Sigma-Aldrich

Monoclonal Anti-SLIPR/MAGI-3 antibody produced in mouse

clone SLP-32, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SLP-32, monoclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1-2 μg/mL using rat brain extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti- SLIPR/MAGI-3 (mouse IgG1 isotype) is derived from the SLP-32 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from mice. Membrane-associated guanylate kinase, WW and PDZ domain-containing 3 (MAGI-3) protein belong to the membrane-associated guanylate kinase (MAGUK) family. This protein is made up of a guanylate-kinase-like region, six post-synaptic density-95/discs large/zonula occludens-1 (PDZ), and two WW domains. MAGUKs localizes to regions of cell-cell contact, such as tight junctions in epithelial cells and synaptic junctions in neurons. The MAGI3 gene is associated with the human chromosome at 1p13.2.

Specificity

Monoclonal Anti-SLIPR/MAGI-3 recognizes rat SLIPR/MAGI-3 (approx. 160 kDa).

Immunogen

C-terminal SLIPR protein fused to GST (amino acids 421-785) of rat SLIPR.

Application

Monoclonal Anti-SLIPR/MAGI-3 antibody produced in mouse may be used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry

Biochem/physiol Actions

Membrane-associated guanylate kinase, WW and PDZ domain-containing 3 (MAGI-3) protein serve to position the phosphatase to specific subcellular locations that are involved in the regulation of cell proliferation and survival. This protein interacts with the protein phosphatase and tensin homolog (PTEN) and receptor protein tyrosine phosphatase β (RPTPβ). MAGUKs are involved in the assembly of multiprotein complexes via their protein-protein interaction domains.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.For prolonged storage, freeze in working aliquots at −20 °C. Repeated freezing and thawing is not recom-mended. Storage in frost-free freezers is also notrecommended. If slight turbidity occurs upon prolongedstorage, clarify the solution by centrifugation beforeuse. Working dilutions should be discarded if not usedwithin 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juncheng Dai et al.
International journal of cancer, 146(10), 2855-2864 (2019-10-03)
Genome-wide association studies (GWAS) have identified 45 susceptibility loci associated with lung cancer. Only less than SNPs, small insertions and deletions (INDELs) are the second most abundant genetic polymorphisms in the human genome. INDELs are highly associated with multiple human
Konstantin Adamsky et al.
Journal of cell science, 116(Pt 7), 1279-1289 (2003-03-05)
Receptor protein tyrosine phosphatase beta (RPTP beta) mediates cell-cell and cell-matrix interactions. By searching for intracellular proteins that interact with the cytoplasmic region of this phosphatase using the two-hybrid method, we identified several proteins containing PDZ domains. One of these
Y Wu et al.
The Journal of biological chemistry, 275(28), 21477-21485 (2000-04-05)
PTEN/MMAC is a phosphatase that is mutated in multiple human tumors. PTEN/MMAC dephosphorylates 3-phosphorylated phosphatidylinositol phosphates that activate AKT/protein kinase B (PKB) kinase activity. AKT/PKB is implicated in the inhibition of apoptosis, and cell lines and tumors with mutated PTEN/MMAC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service