Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S4063

Sigma-Aldrich

SCH-202676 hydrobromide

98%

Synonym(s):

N-(2,3-diphenyl-1,2,4-thiadiazol-5-(2H)-ylidene)methanamine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16BrN3S
CAS Number:
Molecular Weight:
350.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

98%

mp

240.0-240.8 °C (lit.)

solubility

DMSO: 40 mg/mL
H2O: insoluble

SMILES string

Br.C\N=C1\SN(c2ccccc2)C(=N1)c3ccccc3

InChI

1S/C15H13N3S.BrH/c1-16-15-17-14(12-8-4-2-5-9-12)18(19-15)13-10-6-3-7-11-13;/h2-11H,1H3;1H/b16-15+;

InChI key

YJYGOWVFDGULLL-GEEYTBSJSA-N

General description

Modulates radioligand binding to various GPCRs including the human μ-, δ-, and κ-opiod, α- and β-adrenergic, muscarinic M1 and M2, and dopaminergic D1 and D2 receptors.
Shown to modulate radioligand binding to various GPCRs including the human μ-, δ-, and κ-opiod, α- and β-adrenergic, muscarinic M1 and M2, and dopaminergic D1 and D2 receptors. Modulation is G-protein independent and is both selective and reversible.

Application

SCH-202676 hydrobromide may be used in G protein-coupled receptor-mediated cell signaling studies.

Biochem/physiol Actions

Allosteric agonist and antagonist of G-protein coupled receptors (GPCRs).
SCH-202676 inhibits the binding of both agonists and antagonists to G protein-coupled receptors. It also inhibits human μ-, δ-, and κ-opioid, α- and β-adrenergic, muscarinic M1 and M2, and dopaminergic D1 and D2 receptors.[1] Some reports indicates thiol modification as the mechanism of inhibition rather than allosteric modulation.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna M Lewandowicz et al.
British journal of pharmacology, 147(4), 422-429 (2006-01-13)
1. Previous studies suggest that the thiadiazole compound SCH-202676 (N-(2,3-diphenyl-1,2,4-thiadiazol-5-(2H)-ylidene)methanamine) acts as an allosteric modulator of a variety of structurally distinct G protein-coupled receptors (GPCRs). It was postulated that SCH-202676 would directly bind a structural motif in the receptor molecule
A B Fawzi et al.
Molecular pharmacology, 59(1), 30-37 (2000-12-23)
A novel thiadiazole compound, SCH-202676 (N-(2,3-diphenyl-1,2, 4-thiadiazol-5-(2H)-ylidene)methanamine), has been identified as an inhibitor of both agonist and antagonist binding to G protein-coupled receptors (GPCRs). SCH-202676 inhibited radioligand binding to a number of structurally distinct, heterologously expressed GPCRs, including the human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service