Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3826

Sigma-Aldrich

Staphostatin B from Staphylococcus aureus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Staphostatins are a novel group of inhibitors with a unique specificity for the regulation of staphopains, Staphylococcus spp. cysteine proteinases. Staphostatin A and staphostatin B from S. aureus have been characterized in details. Staphostatins interact specifically with this target proteinases forming tight 1:1 and stable non-covalent complex, staphostain A with staphopain A and staphostain B with staphopain B with no cross interaction observed and with very narrow specificity limited to staphopains
The loss of staphostatin B (SspC) expression in S. aureus and the subsequent increase in secreted protease staphopain B (SspB) activity resulted in numerous defects including reduced growth, decreased autolytic activity, resistance to lysis by S. aureus phages, and the inability to secrete proteins into the extracellular matrix. [1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lindsey N Shaw et al.
Journal of bacteriology, 187(5), 1751-1762 (2005-02-18)
The cytoplasmic protein SspC of Staphylococcus aureus, referred to as staphostatin B, is a very specific, tightly binding inhibitor of the secreted protease staphopain B (SspB). SspC is hypothesized to protect intracellular proteins against proteolytic damage by prematurely folded and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service