Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3451

Sigma-Aldrich

SID 852843

≥98% (HPLC)

Synonym(s):

Benzoic acid 5-amino-1-(4-methoxy-benzenesulfonyl)-1H-pyrazol-3-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N3O5S
CAS Number:
Molecular Weight:
373.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)S(=O)(=O)n2nc(OC(=O)c3ccccc3)cc2N

InChI

1S/C17H15N3O5S/c1-24-13-7-9-14(10-8-13)26(22,23)20-15(18)11-16(19-20)25-17(21)12-5-3-2-4-6-12/h2-11H,18H2,1H3

InChI key

FQICFDBMQBCJDT-UHFFFAOYSA-N

Application

SID 852843 may be used in virology studies to inhibit NS3 proteinase of West Nile Virus.

Biochem/physiol Actions

SID 852843 is an inhibitor of West Nile Virus NS2B-NS3 proteinase.
SID 852843 is an inhibitor of West Nile Virus NS2B-NS3 proteinase. Flaviviruses, such as West Nile and Dengue, are a threat to humans from both public health and bioterrorism perspectives. The NS3 proteinase is essential for flaviviral maturation. Inhibitors of NS3 are sought for treatment and prophylactic therapeutics. The NIH molecular library was screened for activity against West Nile Virus NS3, resulting in the discovery of SID 852843. This compound inhibits the NS3 proteinase of West Nile, but not of Dengue type 2 virus. SID 852843 is a reversible, allosteric inhibitor of the formation of the functional conformation of the NS3 proteinase. This is the first flaviviral proteinase inhibitor with the potential to lead to a drug. Aprotinin is an efficient inhibitor of NS3, but it is not selective and has low penetration into cells, limiting its potential for therapeutic use.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service