Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3320

Sigma-Aldrich

Anti-SRRM1 antibody produced in rabbit

~1 mL/mL, affinity isolated antibody, ammonium sulfate suspension

Synonym(s):

Anti-SRm160

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 93.5 kDa

species reactivity

rodent, human

concentration

~1 mL/mL

technique(s)

immunocytochemistry: 1:200
immunohistochemistry (frozen sections): 1:200
western blot: 1:200

UniProt accession no.

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SRRM1(10250)
mouse ... Srrm1(51796)
rat ... Srrm1(313620)

Immunogen

synthetic peptide, PVRRERKRSHSRSPRHR, corresponding to human SRRM1 (SRm160) (amino acids 188-204).

Target description

SRRM1 (SRm160), also referred to as Serine/ArginineRepetitive Matrix Protein 1 and SR-Related NuclearMatrix Protein of 160 kDa, is a member of the SR family of splicing factors. SRm160 functions as a splicing coactivator of pre-mRNA splicing by comple

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service