Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3311

Sigma-Aldrich

SDZ-216,887

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C79H107ClN14O17
Molecular Weight:
1560.23
MDL number:
UNSPSC Code:
12352200

solubility

H2O: >10 mg/mL

storage temp.

−20°C

Amino Acid Sequence

Ac-Nal-Cpa-Trp-Ser-Tyr-Lys-DHP-Leu-Lys-DHP-Pro-Ala-NH2

Application

SDZ-216,887 may be used in LH-RH-mediated cell signaling studies.

Biochem/physiol Actions

SDZ-216,887 blocks the effects of endogenous Luteinizing hormone-releasing hormone (LH-RH) and inhibits the gene expression of pituitary LH-RH receptors.

Features and Benefits

This compound was developed by Sandoz. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

STOT SE 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kovacs et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(4), 1829-1834 (2001-02-15)
The mechanisms through which LH-RH antagonists suppress gonadotroph functions and LH-RH receptor (LH-RH-R) production are incompletely understood. To elucidate these mechanisms, we investigated the effects of Cetrorelix on the mRNA expression of pituitary LH-RH-R and luteinizing hormone (LH) secretion in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service