Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S2687

Sigma-Aldrich

Anti-SDF-1β/PBSF antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Stromal Cell-Derived Factor-1β/Pre-B Cell Growth Stimulating Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCL12(6387)

General description

Stromal cell-derived factor-1 (SDF-1) is a proinflammatory cytokine that belongs to the CXC family of chemokines. SDF-1 exists as two isoforms, SDF-1β and SDF-1α. The effects of SDF-1 are mediated by its binding to the receptor CXCR4, and sometimes to CXCR7. Both lymphoid (bone marrow, spleen and thymus) and non-lymphoid (stomach, kidney, liver, lung, brain, heart) organs express SDF-1. The most important functions assigned to SDF-1 are angiogenesis, migration of μglia and astrocytes, tissue repair and as chemoattractant to hematopoietic cells. Studies also implicate SDF-1 in survival of human glioma cells via the activation of PI3K/Akt pathway. The other pathways that are activated by SDF-1 include MAPK, NF-κB, paxillin, Crk and other focal adhesion proteins. A variety of human malignancies of pancreas, breast, prostate and glioma exhibit overexpression of CXCR4. High expression of SDF-1 has been observed in lymph nodes, lungs, bone and liver that are the most common sites of metastasis of breast cancer cells. Anti- SDF-1β/PBSF recognizes recombinant human SDF-1β/PBSF. By immunoblotting, this antibody exhibits less than 5% cross-reactivity with recombinant human SDF-1α.

Immunogen

recombinant human SDF-1β/PBSF expressed in E. coli.

Application

Anti-SDF-1β/PBSF antibody may be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml. For ELISA, the recommended working antibody concentration is 0.5-1.0 μg/ml. The antibody is also suitable for neutralization assays; ND50 of the antibody is ~5-15 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R K Ganju et al.
The Journal of biological chemistry, 273(36), 23169-23175 (1998-08-29)
The alpha-chemokine stromal cell-derived factor (SDF)-1alpha binds to the seven transmembrane G-protein-coupled CXCR-4 receptor and acts to modulate cell migration and proliferation. The signaling pathways that mediate the effects of SDF-1alpha are not well characterized. We studied events following SDF-1alpha
T Nagasawa et al.
Seminars in immunology, 10(3), 179-185 (1998-07-07)
PBSF/SDF-1 is a CXC chemokine which has unique functions among chemokines. It is essential for viability of the embryo, B lymphopoiesis, bone marrow hematopoiesis and cardiogenesis. A receptor for PBSF/SDF-1 was shown to be CXCR4 that is an entry co-receptor
D Tseng et al.
British journal of cancer, 104(12), 1805-1809 (2011-05-19)
Local recurrence of glioblastomas is a major cause of patient mortality after definitive treatment. This review discusses the roles of the chemokine stromal cell-derived factor-1 and its receptor CXC chemokine receptor 4 (CXCR4) in affecting the sensitivity of glioblastomas to
Yan Zhou et al.
The Journal of biological chemistry, 277(51), 49481-49487 (2002-10-22)
Chemokines were described originally in the context of providing migrational cues for leukocytes. They are now known to have broader activities, including those that favor tumor growth. We addressed whether and which chemokines may be important promoters of the growth
A Müller et al.
Nature, 410(6824), 50-56 (2001-03-10)
Breast cancer is characterized by a distinct metastatic pattern involving the regional lymph nodes, bone marrow, lung and liver. Tumour cell migration and metastasis share many similarities with leukocyte trafficking, which is critically regulated by chemokines and their receptors. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service