Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S2565

Sigma-Aldrich

Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit

affinity isolated antibody, aqueous glycerol solution

Synonym(s):

Anti-STAT1(pT701)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

aqueous glycerol solution

species reactivity

mouse, human (weaker), rat

technique(s)

western blot: 1:500 using mouse 3T3-LI cells or rat adipocytes exposed to LIF, oncostatin M, IFN-γ, or growth hormone

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT1(6772)
mouse ... Stat1(20846)
rat ... Stat1(25124)

Immunogen

synthetic phosphopeptide derived from the region of human STAT 1 that contains tyrosine701.

Physical form

Solution in Tris-buffered saline, pH 7.4, containing 50% glycerol, 1 mg/ml BSA (IgG and protease free) and 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service