Streptavidin iron oxide particles are used in protein chromatography, affinity chromatography, and recombinant protein expression and analysis. Streptavidin iron oxide particles have been used to study imaging of cellular spread on a three-dimensional scaffold by means of a novel cell-labeling technique for high-resolution computed tomography. Streptavidin iron oxide particles have also been used to study in vitro biomedical applications of functionalized iron oxide nanoparticles.
Used for the purification of biotin containing proteins or DNA binding proteins
Physical form
Suspension in 0.85% NaCl, 0.01 M phosphate, pH 8.0, containing 0.1% bovine serum albumin, 0.1% sodium azide
Journal of the American Chemical Society, 127(9), 3165-3171 (2005-03-03)
Aptamers are DNA (or RNA) ligands selected from large libraries of random DNA sequences and capable of binding different classes of targets with high affinity and selectivity. Both the chances for the aptamer to be selected and the quality of
Contrast media & molecular imaging, 6(4), 236-250 (2011-08-24)
Superparamagnetic iron oxide nanoparticles (SPION) are very promising contrast media, especially for molecular imaging, due to their superior NMR efficacy. They even have wider biomedical applications such as in drug and gene delivery, tissue engineering and bioseparation, or as sensitive
The EMBO journal, 39(3), e102500-e102500 (2019-12-17)
RNase J1 is the major 5'-to-3' bacterial exoribonuclease. We demonstrate that in its absence, RNA polymerases (RNAPs) are redistributed on DNA, with increased RNAP occupancy on some genes without a parallel increase in transcriptional output. This suggests that some of
Tissue engineering. Part C, Methods, 18(3), 167-175 (2011-09-29)
Computed tomography (CT) represents a truly three-dimensional (3D) imaging technique that can provide high-resolution images on the cellular level. Thus, one approach to detect single cells is X-ray absorption-based CT, where cells are labeled with a dense, opaque material providing
Journal of immunological methods, 458, 26-32 (2018-04-15)
Hepatitis B virus (HBV) infection is the major public health problem leading cause of death worldwide. The most important diagnostic marker for this infection is hepatitis B surface antigen (HBsAg). In this study, a novel, inexpensive, portable and sensitive ELISA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.