Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S2195

Sigma-Aldrich

Anti-SFRS1/9 antibody produced in rabbit

affinity isolated antibody, ammonium sulfate suspension

Synonym(s):

Anti-Splicing Factor Arginine/serine-rich 1/9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 27.7 kDa

species reactivity

human, rodent

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFRS1(6426)
mouse ... Sfrs1(110809)
rat ... Psip1(313323)

Immunogen

synthetic peptide corresponding to human SFRS1/9 (LDNTKFRSHEGET) conjugated to BSA.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuxi Wang et al.
The FEBS journal, 283(1), 102-111 (2015-10-16)
Microtubules are dynamic assemblies of αβ-tubulin heterodimers and have been recognized as highly attractive targets for cancer chemotherapy. A broad range of agents bind to tubulin and interfere with microtubule assembly. Despite having a long history of characterization, colchicine binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service