Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1943

Sigma-Aldrich

Serotonin Transporter Receptor

membrane suspension

Synonym(s):

Serotonin Transporter Receptor human from human platelets, SERT, 5-HT transporter, 5-HTT

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

form

membrane suspension

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SLC6A4(6532)

General description

Serotonin Transporter Receptor Preparation, human is a frozen aliquot of membranes from human platelets suitable for binding assays. Standard binding assay protocol can be found on the product datasheet and specific binding capacity is provided for each lot.

Physical form

Membranes suspended in 50 mM TRIS-HCL containing 120 mM NaCl and 5 mM KCl, pH 7.4 Actual concentration and specific binding capacity are provided with each lot.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service