Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1757

Sigma-Aldrich

Sera canine

lyophilized powder (from clotted whole blood)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

biological source

canine

description

Contains no azides

form

lyophilized powder (from clotted whole blood)

storage temp.

2-8°C

Application

Canine serum is used to support the in vitro cultivation of canine protozoan parasites such as Babesia canis canis and Babesia gibsoni. Not tested for use in cell culture.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro cultivation of Babesia canis canis parasites isolated from dogs in Poland.
Adaszek L, Winiarczyk S
Parasitology Research (2010)
E Zweygarth et al.
Parasitology research, 86(11), 905-907 (2000-11-30)
Two stocks of the protozoan parasite Babesia gibsoni, one of the causative agents of canine piroplasmosis, were propagated continuously in dog erythrocytes in microaerophilous stationary-phase culture. Cultures of both stocks were initiated in a humidified 5% CO2, 2% O2, 93%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service