By immunoblotting, the antibody reacts specifically with SHIP (145 kDa and/or 135 kDa) as well as C-terminal truncated forms (125 kDa and 110 kDa) in some cell types.
Immunogen
GST fusion protein corresponding to residues 7-133 of murine SHIP (Src homology 2-containing inositol phosphatase) SH2 domain.
Physical form
Solution in 0.07 M Tris-glycine, pH 7.4, containing 0.105 M sodium chloride, 30% glycerol and 0.035% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.