Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

S1072

Sigma-Aldrich

Anti-Sodium Channel ASIC4 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ACCN4, Anti-H+-gated cation channel, Anti-SPASIC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200-1:500 using rat brain lysate, ND7/23 cell line lysate, and rat dorsal root ganglion (DRG) lysate

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

peptide CKIKFAEEDAKPKEKEAGDE corresponding to residues 7-26 of N-terminal rat Accn4.

Target description

Sodium Channel ASIC4 is a member of the ASIC family that also includes ASIC1, ASIC2 and ASIC3. The ASIC family is part of a large channel superfamily known as degenerin/epithelial sodium channels (DEG/ENaC). The functional ASIC channel is composed of 4 subunits, which can form either a homo or heterotetramer. ASIC4 protein does not appear to function as a proton-gated channel when expressed alone in heterologous systems and therefore a modulatory role for this subunit has been proposed. ASIC4 protein is expressed in brain, pituitary gland, spinal cord and inner ear.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amra Saric et al.
Molecular biology of the cell, 27(2), 321-333 (2015-11-20)
Macrophages and dendritic cells exposed to lipopolysaccharide (LPS) convert their lysosomes from small, punctate organelles into a network of tubules. Tubular lysosomes have been implicated in phagosome maturation, retention of fluid phase, and antigen presentation. There is a growing appreciation
Blandine Monel et al.
The EMBO journal, 36(12), 1653-1668 (2017-05-06)
The cytopathic effects of Zika virus (ZIKV) are poorly characterized. Innate immunity controls ZIKV infection and disease in most infected patients through mechanisms that remain to be understood. Here, we studied the morphological cellular changes induced by ZIKV and addressed
Priya Crosby et al.
Cell, 177(4), 896-909 (2019-04-30)
In mammals, endogenous circadian clocks sense and respond to daily feeding and lighting cues, adjusting internal ∼24 h rhythms to resonate with, and anticipate, external cycles of day and night. The mechanism underlying circadian entrainment to feeding time is critical for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service