S1064
Monoclonal Anti-SERCa1 ATPase antibody produced in mouse
clone IIH11, ascites fluid
Synonym(s):
Anti-Sarcoplasmic or Endoplasmic Reticulum Ca2+ ATPase1
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
IIH11, monoclonal
contains
0.05% sodium azide
species reactivity
rat, rabbit, canine, guinea pig, mouse
should not react with
human
technique(s)
indirect immunofluorescence: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
mouse ... Atp2a1(11937)
rat ... Atp2a1(116601)
Immunogen
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service