Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

S102

Sigma-Aldrich

S-(−)-SKF-38393 hydrochloride

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO2 · HCl
CAS Number:
Molecular Weight:
291.77
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

optical activity

[α]27/D −14.8°, c = 1.7 in methanol(lit.)

color

tan

SMILES string

Cl.Oc1cc2CCNC[C@@H](c3ccccc3)c2cc1O

InChI

1S/C16H17NO2.ClH/c18-15-8-12-6-7-17-10-14(13(12)9-16(15)19)11-4-2-1-3-5-11;/h1-5,8-9,14,17-19H,6-7,10H2;1H/t14-;/m0./s1

Inchi Key

YEWHJCLOUYPAOH-UQKRIMTDSA-N

Biochem/physiol Actions

D1 dopamine receptor agonist; less active enantiomer of (±)-SKF-38393.

Caution

Photosensitive

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gnana P Krishnamoorthy et al.
Cancer discovery, 9(2), 264-281 (2018-10-12)
Translation initiation is orchestrated by the cap binding and 43S preinitiation complexes (PIC). Eukaryotic initiation factor 1A (EIF1A) is essential for recruitment of the ternary complex and for assembling the 43S PIC. Recurrent EIF1AX mutations in papillary thyroid cancers are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service