Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0818

Sigma-Aldrich

Anti-Secretory Carrier Membrane Protein 2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SCAMP 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~38 kDa

species reactivity

hamster, mouse

technique(s)

western blot: 2 μg/mL using CHO cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SCAMP2(10066)
mouse ... Scamp2(24044)

Specificity

Detects secretory carrier membrane protein 2 (SCAMP 2) from mouse testes and AtT20 cell extract as well as Chinese hamster ovary (CHO) cell extract.

Immunogen

synthetic peptide corresponding to amino acid resudues 1-17 from human SCAMP 2. This sequence is completely conserved between human and mouse.

Physical form

Solution in phosphate buffered saline containing 1 mg/ml bovine serum albumin and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucile Poisson et al.
Scientific reports, 10(1), 13350-13350 (2020-08-10)
The acquisition of invasive capacities by carcinoma cells, i.e. their ability to migrate through and to remodel extracellular matrices, is a determinant process leading to their dissemination and to the development of metastases. these cancer cell properties have often been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service