S0810
Silicone isolators, Press-to-Seal
1 well, D × L × W 1.0 mm × 32 mm × 19 mm, silicone/adhesive, oblong
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
MA.01
Recommended Products
material
oblong
silicone/adhesive
sterility
non-sterile
manufacturer/tradename
Grace Bio-Labs
D × L × W
1.0 mm × 32 mm × 19 mm
Application
Isolate specimens on a microscope slide with hydrophobic press-to-seal (removable) or peel-and-stick (adhesive) barriers. Available in silicone thickness 0.5 to 2.5mm as pre-cut gaskets and sheet material; with or without secure seal clean release adhesive on one or both surfaces. Non-adhesive isolators are autoclavable and re-usable for cell culture applications. NOTE: Add approximately 0.15mm to depth per layer of adhesive.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Janosch Peter Heller et al.
Methods (San Diego, Calif.), 174, 81-90 (2019-06-04)
Astroglia are vital facilitators of brain development, homeostasis, and metabolic support. In addition, they are also essential to the formation and regulation of synaptic circuits. Due to the extraordinary complex, nanoscopic morphology of astrocytes, the underlying cellular mechanisms have been