microarray: suitable western blot: 1.5-3.0 μg/mL using whole cell extract of mouse fibroblast NIH3T3 cell line western blot: 3-5 μg/mL using mouse brain and rat brain extract (S1 fraction)
Striatin (STRN) is also termed as calmodulin-binding protein. Heart, kidney, several digestive organs and the adrenal shows its expression. STRN is located on human chromosome 2p22.2. It is a neuronal, membrane associated protein. Striatin is multimodular and belongs to a family of Ca2+ and calmodulin binding proteins with WD motif repeats, that includes SG2NA and zinedin. Striatin is found in low abundance in the rat brain. It is mostly expressed in neurons of the motor and olfactory structures of the central nervous system, predominantly in the striatum. It is found to be expressed in the somatodendritic compartment of neurons and in dendritic spines.
Specificity
Anti-Striatin recognizes mouse and rat striatin, 110 kDa.
Immunogen
synthetic peptide corresponding to amino acids 378-396 located in the mid region of mouse striatin, conjugated to KLH. This sequence is highly conserved in rat and human striatin (84-89% identity).
Application
Anti-Striatin antibody produced in rabbit may be used in immunoblotting.
Biochem/physiol Actions
Striatin (STRN) forms a complex with protein phosphatase 2A (PP2A) and has been shown to function as an estrogen receptor alpha (ERα)-binding protein. It plays a key role as a molecular anchor which helps to localize ERα to the membrane. STRN also aid in systematizing the ERα-endothelial NOS (eNOS) membrane signaling complex. It has been suggested that striatin is an adaptor or transducer protein involved in Ca2+ and calmodulin-dependent signaling events within dendritic spines. Down regulation of striatin, which is expressed in a few subsets of neurons, impairs the growth of dendrites as well as locomotor activity.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Storage and Stability
For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Detection of an ALK fusion in colorectal carcinoma by hybrid capture-based assay of circulating tumor DNA
Lai AZ, et al.
Oncologist, 22(7), 774-774 (2017)
Striatin assembles a membrane signaling complex necessary for rapid, nongenomic activation of endothelial NO synthase by estrogen receptor alpha
Lu Q, et al.
Proceedings of the National Academy of Sciences, 101(49), 17126-17131 (2004)
Interaction of calmodulin with striatin, a WD-repeat protein present in neuronal dendritic spines
Bartoli M, et al.
Test, 273(35), 22248-22253 (1998)
Striatin gene polymorphic variants are associated with salt sensitive blood pressure in normotensives and hypertensives
Gupta T, et al.
American Journal of Hypertension, 31(1), 124-131 (2018)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.